missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ G7000HHR HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021895
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021895 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021895 Supplier Merck Emd Millipore Supplier No. 817359

TSKgel G7000HHR Columns are packed with rigid, porous, polymer beads. They are used for the non-aqueous separation of polymers with estimated molecular weights less than 400,000,000 Da.

  • Conventional GPC columns with dimensions of 7.8 mm ID x 30 cm
  • All TSKgel HHR-type columns are shipped in THF, which can be exchanged for a broad range of organic solvents
  • Maximum Operating Temperature: 140°C

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 2 mL/min
Length (Metric) 30 cm
Max. Pressure 50 bar
Particle Size 5 μm
Temperature 140°C
USP Type L21
Stationary Phase Polystyrene-divinylbenzene
Product Line TSKgel
Shipping Condition Shipped in THF
Matrix Polystyrene-divinylbenzene Phase/Polymer Particle Platform
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation