missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ TSKgel™ G3000SWXL HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021808
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021808 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021808 Supplier MilliporeSigma™ Supelco™ Supplier No. 808541

Widely considered the premium columns for analyzing biopolymers by GFC, TSKgel™ SWXL columns feature high pore volume per unit column volume, low sample adsorption and excellent column efficiency, all contributing to unsurpassed sample resolution.

TSKgel™ G3000SWXL columns are an excellent choice for protein and large peptide separations. The fractionation range is applicable for globular protein samples up to 0.5 million Da.

Order Info

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Specifications

Column Format HPLC Column
Diameter (Metric) 7.8 mm
Flow Rate 1.2 mL/min.
Length (Metric) 30 cm
Max. Pressure 72 bar
Particle Shape Silica Particle Platform
Particle Size 5 μm
pH 2.5 to 7.5
Pore Size 250 Å
Solvents 0 to 100% aqueous soluble organic solvents
Temperature 10°C to 30°C
USP Type L20, L59
Stationary Phase Diol
Product Line TSKgel
Matrix Silica Particle Platform
Packing Material Silica Particle Platform
Column Type Size Exclusion
Phase Size Exclusion
Material Stainless Steel
Technique HPLC
Product Type HPLC Column
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation