missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKgel™ G2000SW HPLC Column, 13 μm Particle Size, Sold by MilliporeSigma™ Supelco™
TSKgel™ G2000SW SEC columns provide excellent separation of peptides and proteins with molecular weights up to 100,000 Da.
$9041.43 - $13164.46
Specifications
| Diameter (Metric) | 21.5 mm |
|---|---|
| Flow Rate | 8 mL/min. |
| Particle Size | 13 μm |
| pH | 2.5 - 7.5 |
| Pore Size | 125 Å |
| Catalog Number | Mfr. No. | Length (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (Metric) | Price | Quantity | |||||
|
111021753
|
Merck Emd Millipore
806727 |
30 cm |
Each for $9,041.43
|
|
|||||
|
111021739
|
Merck Emd Millipore
805146 |
60 cm |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
TSKgel™ G2000SW columns start at 10 μm for analytical columns and also scale to 13 and 20 μm in columns up to 55 mm I.D. for preparative purifications.Order Info
Shipped in 0.05% NaN3 and 0.1 mol/L Na2SO4 in 0.1 M phosphate buffer, pH 6.7.
Specifications
| 21.5 mm | |
| 13 μm | |
| 125 Å | |
| 10°C to 30°C | |
| Diol | |
| Silica Particle Platform | |
| Stainless Steel Hardware |
| 8 mL/min. | |
| 2.5 - 7.5 | |
| 0 to 100% aqueous soluble organic solvents | |
| L20, L59 | |
| TSKgel | |
| Size Exclusion |
TSKgel is a registered trademark of Tosoh Corporation