missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ G2000HXL HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021864
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021864 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021864 Supplier Merck Emd Millipore Supplier No. 816134
Only null left

TSKgel G2000HXL Columns are packed with rigid, porous, polymer beads. They are applied to the non-aqueous separation of polymers with molecular weights of less than 10,000 Da.

  • Use TSKgel Hhr-type columns when a broader range of shipping solvents is needed
  • THF can be replaced with benzene, chloroform, toluene, xylene, dichloromethane, and dichloroethane

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1.2 mL/min
Length (Metric) 30 cm
Max. Pressure 50 bar
Particle Size 5 μm
Pore Size 20 Å
Temperature 60°C
Stationary Phase Polystyrene-divinylbenzene
Product Line TSKgel
Shipping Condition Shipped in THF
Matrix Polystyrene-divinylbenzene Phase/Polymer Particle Platform
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation