missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ Ether-5PW HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021973
Change view
Click to view available options
Diameter (Metric):
2 mm
5 mm
7.5 mm
8 mm
Material:
Glass
Stainless Steel
Stainless Steel Hardware
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Material
111021973 2 mm Stainless Steel Hardware
111021844 5 mm Glass
111021845 8 mm Glass
111021814 7.5 mm Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111021973 Supplier Merck Emd Millipore Supplier No. 818760

An excellent choice for separating hydrophobic molecules such as membrane proteins, or monoclonal antibodies such as IgG or IgM.

  • Among the TSKgel HIC Columns, TSKgel Ether-5PW has intermediate hydrophobity
  • An excellent choice for separating hydrophobic molecules such as membrane proteins or monoclonal antibodies such as IgG or IgM
  • Features a 1000 Å pore size in a polymethacrylate base material
  • Column is stable in either acid or caustic cleaning regimens
  • Provides excellent access to larger molecules with low diffusion coefficients
  • Both hydrophobic interaction chromatography (HIC) and reversed-phase liquid chromatography separate on the basis of protein hydrophobicity allowing for selective binding, and desorption of proteins
  • Recommended for purifying more hydrophobic proteins
  • Salt Concentration: ≤4 M

Specifications

Diameter (Metric) 2 mm
Flow Rate 0.12 mL/min
Length (Metric) 7.5 cm
Max. Pressure 6 bar
Particle Size 10 μm
pH 2 to 12
Pore Size 1000 Å
Solvents ≤ 50% Organic Solvent
Temperature 10°C to 50°C
Stationary Phase Ether
Product Line TSKgel
Matrix Ether Phase/Hydroxylated Methacrylic Polymer
Material Stainless Steel Hardware
Technique HIC HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation