missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ DEAE-SW Columns, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021974
Change view
Click to view available options
Length (Metric):
25 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021974 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021974 Supplier Merck Emd Millipore Supplier No. 818761

TSKgel DEAE-SW Columns are packed with 5 μm porous, spherical silica beads which are surface modified with a weak anion exchanger. They are used for the separation and analysis of nucleotides, nucleosides, peptides, small proteins, and other biologically active molecules.

  • A 2 mm ID column is available for applications that are sample limited

Specifications

Diameter (Metric) 2 mm
Flow Rate 0.22 mL/min
Ion Exchange Group H2PO4-
Length (Metric) 25 cm
Max. Pressure 130 bar
Particle Size 5 μm
pH 2 to 7.5
Pore Size 125 Å
Solvents 0-100% Organic Solvent
Temperature 10°C to 45°C
Stationary Phase DEAE (diethylaminoethyl)
Product Line TSKgel
Matrix DEAE (diethylaminoethyl) Phase/Silica Particle Platform
Material Stainless Steel Hardware
Technique Anion Exchange
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation