missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ CN-80Ts HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021888
Change view
Click to view available options
Length (Metric):
15 cm
25 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021888 25 cm
111021887 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111021888 Supplier Merck Emd Millipore Supplier No. 817349

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Comprised of cyanopropyl groups bonded to the silica. Applications include reversed-phase high performance liquid chromatographic techniques.

  • An alternative to C18 (ODS) and C8 (Octyl) phases
  • The cyano group is the least hydrophobic of the 100 Å phases available and in some cases is used under normal phase conditions
  • Useful for the analysis of polar peptides, amino acids, and other pharmaceutical and food & beverage products
  • Provides reproducible separations of molecules below 6,000 MW
  • Shipped in 60% Acetonitrile, 40% Water

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1.2 mL/min
Length (Metric) 25 cm
Max. Pressure 300 bar
Particle Size 5 μm
pH 2 to 7.5
Pore Size 80 Å
Solvents 0-100% Organic Solvent
Surface Area 425 m2/g
Temperature 10°C to 50°C
USP Type L10
Stationary Phase Cyano
Product Line TSKgel
Shipping Condition Shipped in 60% Acetonitrile, 40% Water
Matrix Cyano Phase/Silica Particle Platform
Carbon Load 8%
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel Hardware
Technique Reverse Phase HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation