missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKgel™ Chelate-5PW HPLC Column, Sold by MilliporeSigma™ Supelco™
Description
- High efficiency, resin-based affinity columns separate or purify many enzymes and other proteins
- Utilize the tendency of iminodiacetic acid (IDA) to form a complex with metal ions, such as Zn2+, Ni2+ and Cu2+
- Column is loaded with divalent metal ions by chelation
- Peptides and proteins containing histidine residues will normally adsorb to these chelated ions at a neutral pH
- Retained compounds can be eluted with buffer containing imidazole or glycine
- A decreasing pH gradient can be used for elution purposes as well
- For successful use of this retention mechanism, select the proper metal ions for chelation and an elution buffer to desorb the analytes
- Applications: immunoglobulins, transferrin, lectins, milk proteins, membrane proteins and peptides
Specifications
Specifications
| Diameter (Metric) | 5 mm |
| Flow Rate | 1 mL/min |
| Length (Metric) | 5 cm |
| Max. Pressure | 20 bar |
| Particle Size | 10 μm |
| pH | 2 to 12 |
| Pore Size | 1000 Å |
| Solvents | ≤ 20% Organic Solvent |
| Temperature | ≤45°C |
| Stationary Phase | Iminodiacetic Acid |
| Show More |
TSKgel is a registered trademark of Tosoh Corporation