missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ BioAssist™ S HPLC Column, 13 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022042
Change view
Click to view available options
Length (Metric):
10 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111022042 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022042 Supplier Merck Emd Millipore Supplier No. 821411

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Packed with porous, hydrophilic polymer beads that are surface modified with a strong cation exchanger.

  • Packed with spherical, polymer beads (1300 Å)
  • Surface modified with strong sulfopropyl cation exchange groups
  • Large pores allow large biomolecules to freely enter and exit the pores
  • Offers higher binding capacity and better recovery of activity than other competivie materials
  • Standard column dimensions are 4.6 mm ID x 5 cm
  • A 10 mm ID x 10 cm semi-preparative column is available for scale-up
  • Columns are made of PEEK to reduce protein adsorption
  • Salt Concentration: ≤ 2.6 M

Specifications

Diameter (Metric) 10 mm
Flow Rate 5 mL/min
Ion Exchange Group Na+
Length (Metric) 10 cm
Max. Pressure 25 bar
Particle Size 13 μm
pH 2 to 12 (≤ 1 Month), 3 to 10 (> 1 Month)
Pore Size 1300 Å
Solvents ≤ 30% Organic Solvent
Temperature 4°C to 60°C
Stationary Phase Sulfonic Acid
Product Line TSKgel
Shipping Condition Shipped in 20% Ethanol in 20 mM Phosphate Buffer, pH 6.5
Matrix Sulfonic Acid Phase/Polymer Particle Platform
Material PEEK Hardware
Technique Cation Exchange
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation