missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ Amide-80 HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021853
Change view
Click to view available options
Diameter (Metric):
7.8 mm
21.5 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
111021853 7.8 mm
111021854 21.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111021853 Supplier Merck Emd Millipore Supplier No. 814459

For the analysis of saccharides, glycans, oligosaccarides, peptides, and polar compounds from natural products.

  • Columns are packed with 3, 5, or 10 micron spherical silica particles covalently bonded with carbamoyl groups
  • Amide stationary phase provides a unique selectivity under regular normal phase conditions, or in the hydrophilic interaction mode of chromatography
  • Target applications include the analysis of saccharides, glycans, oligosaccarides, peptides, and polar compounds from natural products
  • Unique advantages of Amide-80 phase for saccharide analysis include a novel hydrogen bonding retention mechanism between hydroxyl groups of the sample and the carbamoyl group in the stationary phase
  • Stationary phase does not react with reducing sugars
  • Can be used at elevated temperatures (≤ 80°C) to prevent peak splitting of carbohydrates
  • High efficiency, 3 micron columns offer shorter analysis time and enhanced sensitivity for LC-MS applications
  • Shipped in 85% acetonitrile, 15% water

Specifications

Diameter (Metric) 7.8 mm
Length (Metric) 30 cm
Max. Pressure 70 bar
Particle Shape Sphere
Particle Size 10 μm
pH 2 to 7.5
Pore Size 100 Å
Solvents 0-100% Organic Solvent
Surface Area 450 m2/g
Temperature 10°C to 80°C
USP Type L68
Stationary Phase Carbamoyl
Product Line TSKgel
Shipping Condition Shipped in 85% Acetonitrile, 15% Water
Matrix Carbamoyl Phase/Silica Particle Platform
Material Stainless Steel Hardware
Technique HILIC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation