missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ Alpha-M HPLC Column, 13 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021944
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021944 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021944 Supplier Merck Emd Millipore Supplier No. SGMA818344

TSKgel Alpha-M Columns are so-called mixed-bed columns with an extended linear calibration range. They are packed with rigid, porous, polymer beads which show minimal shrinkage or swelling in different solvent systems.

  • Use for the separation of polymers that are soluble in polar organic solvents such as methanol, ethanol, DMF, THF, HFIP, DMSO, 2-propanol, and acetonitrile
  • TSKgel Alpha columns are composed of spherical, hydrophilic methacrylate beads
  • Stable in a wide variety of organic solvents at concentrations up to 100%.
  • Suitable for samples with a broad MW distribution, as well as samples with unknown molecular weight
  • Salt Concentration: ≤ 0.5 M

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1 mL/min
Length (Metric) 30 cm
Max. Pressure 20 bar
Particle Size 13 μm
pH 2 to 12
Solvents 0-100% Polar Organic Solvent
Temperature 80°C
USP Type L38, L39
Stationary Phase Hydroxylated Methacrylate
Product Line TSKgel
Shipping Condition Shipped aq. soln.
Matrix Hydroxylated Methacrylate Phase/Polymer Particle Platform
Material Stainless Steel Hardware
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation