missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Trypsin, Porcine Pancreas

Catalog No. ICN15357125
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15357125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15357125 Supplier MP Biomedicals™ Supplier No. 0215357125
Only null left

Typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.

  • CAS Number: 9002-07-7
  • Molecular Weight: Approximately 23,000 or 22000 (for human)
  • Composition: Trypsin is composed of two subunits, a-trypsin and b-trypsin. a-Trypsin is composed of two peptide chains and b-trypsin is composed of one chain.
  • Activity: One BAEE unit will produce a DA253 of 0.001 per minute at pH 7.6 at 25°C using BAEE as substrate. Reaction volume = 3.2mL (1 cm light path).
    One TAME unit hydrolyzes 1 umole of p-toluene-sulfonyl-L-arginine methyl ester (TAME) per minute at 25°C, pH 8.2, in the presence of 0.001 M calcium ion.
    One USP trypsin unit is the activity causing a change in absorbance of 0.003 per minute under the conditions specified.
  • Activity Conversion:1 TAME unit = 19.2 USP or NF units = 57.5 BAEE Units
  • Composition: Trypsin is composed of two subunits, a-trypsin and b-trypsin. a-Trypsin is composed of two peptide chains and b-trypsin is composed of one chain.

Specifications

For Use With (Application) Release adherent cells from tissue culture plates for passaging
Sample Type Porcine
Content And Storage 4°C
Form Powder
Quantity 25 g
Product Type Trypsin