missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TrueMark™ Respiratory Panel 2.0 PreAmp Primers

Catalog No. A49049
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A49049 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A49049 Supplier Applied Biosystems™ Supplier No. A49049

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The TrueMark Respiratory Panel 2.0 PreAmp Primers kit contains primers for all respiratory tract microbiota assays plus the TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control and the RNase P RPPH1 gene, but does not contain primers for the TaqMan Universal Extraction Control Organism (B. atrophaeus).

The TrueMark Respiratory Panel 2.0 PreAmp Primers are intended to be used in the preamplification step of the TrueMark Respiratory Panel 2.0, OpenArray plate and TrueMark Respiratory Panel 2.0, TaqMan array card workflows to help achieve the highest analytical sensitivity with the added benefit of sample conservation.

Specifications

Content And Storage Store at -30°C to -5°C
Detection Method Primer-probe
For Use With TrueMark™ Respiratory Panel 2.0, TaqMan™ Array Card, TrueMark™ Respiratory Panel 2.0, OpenArray™ Plate
Product Line TrueMark
Quantity 1 Kit
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.