missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TrueMark™ Respiratory Panel 2.0 Amplification Control

Catalog No. A48101
Encompass
Change view
Click to view available options
Quantity:
5 x 50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A48101 5 x 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A48101 Supplier Applied Biosystems™ Supplier No. A48101
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The TrueMark Respiratory Panel 2.0 Amplification Control contains a linearized multi-target plasmid with target sequences for each respiratory tract microbiota assay on the TrueMark Respiratory Panel 2.0, OpenArray plate and TrueMark Respiratory Panel 2.0, TaqMan array card.

The TrueMark Respiratory Panel 2.0 Amplification Control can be included in respiratory tract microbiota profiling experiments as a positive control for panel-specific amplification.

The plasmid also contains target sequences for TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control, TaqMan Universal Extraction Control Organism (B. atrophaeus), and the human RNase P RPPH1 gene. In addition, the plasmid contains target sequences for additional qualified TaqMan assays for respiratory tract microbiota profiling that may be included on custom arrays or cards. Refer to the Respiratory Tract Microbiota Profiling Experiments v2 using TaqMan Array Cards Application Guide or Respiratory Tract Microbiota Profiling Experiments v2 using OpenArray Application Guide for the full assay list.

The TrueMark Respiratory Panel 2.0 Amplification Control is supplied at a concentration of 1 x 105 copies/μL. During real-time PCR, the control can be used as a stand-alone sample to verify assay performance and to help with troubleshooting.

Specifications

Concentration 1 x 105 Copies/μL
Content And Storage Store at -25°C to -15°C.
Detection Method Primer-probe
Format Tube
For Use With TrueMark™ Respiratory Panel 2.0, TaqMan™ Array Card, TrueMark™ Respiratory Panel 2.0, OpenArray™ Plate
Product Line TrueMark
Quantity 5 x 50 μL
Shipping Condition Dry Ice
Volume 250 μL

For Research Use Only. Not for use in diagnostic procedures.