missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TrueMark™ Respiratory II Plus Combo Kit

Catalog No. A56286C
Encompass
Change view
Click to view available options
5 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A56286C 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A56286C Supplier Applied Biosystems™ Supplier No. A56286C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems TrueMark Respiratory II Plus Combo Kit achieves molecular detection through real-time PCR technology, offering an end-to-end workflow for many common infectious disease areas by utilizing the power of QuantStudio real-time PCR systems.

The Applied Biosystems TrueMark Respiratory II Plus Combo Kit achieves molecular detection through real-time PCR technology, offering an end-to-end workflow for many common infectious disease areas by utilizing the power of QuantStudio real-time PCR systems. The combination of a TrueMark panel (included) and a QuantStudio system helps enable rapid, accurate detection and categorization of microorganisms that cause respiratory diseases.

The TrueMark Respiratory II Plus panel is composed of convenient, duplexed TaqMan assays that detect coronavirus 229E, coronavirus HKU1, coronavirus NL63, coronavirus OC43, parainfluenza virus 1, parainfluenza virus 2, parainfluenza virus 3, parainfluenza virus 4, and an internal control in the same reaction, utilizing real-time PCR techniques.

Features of the TrueMark Respiratory II Plus panel include:
• Ease-of-use—convenient, duplexed reactions include assay and positive control, pre-spotted and dried down on TaqMan plates, designed for easy setup and increased accuracy; just add sample and TrueMark Infectious Disease Panels 1-Step Multiplex Master Mix (No ROX) (included)
• Endogenous controls—duplex reactions offer RNase P human internal control in every well for added confidence
• Turn-around time—choose from a selection of predefined panels with a short turn-around time
• Amplification control—TrueMark Amplification Control I covers all targets in RTI, UTI, STI, and vaginal panels
• Maximize the workflow—expand from SARS-CoV-2 testing to TrueMark infectious disease research panels utilizing the same workflow

The TrueMark Respiratory II Plus Combo Kit includes the following components:
• TrueMark Respiratory II Plus Panel—collection of duplex assays that contain primer/probe sets specific to coronavirus 229E, coronavirus HKU1, coronavirus NL63, coronavirus OC43, parainfluenza virus 1, parainfluenza virus 2, parainfluenza virus 3, parainfluenza virus 4, and RNase P genomic regions
• TrueMark Infectious Disease Panels 1-Step Multiplex Master Mix (No ROX)—ready-to-use PCR mix, including reverse transcriptase, polymerase, dNTPs, salt, and buffer

The TrueMark Respiratory II Plus Combo Kit is used with nasopharyngeal swab or nasopharyngeal aspirate samples on our QuantStudio real-time PCR systems.

Product use
The TrueMark Respiratory II Plus Panel is a collection of duplex assays that detect microbes and an internal control in the same reaction for qualitative detection and identification of coronavirus 229E, coronavirus HKU1, coronavirus NL63, coronavirus OC43, parainfluenza virus 1, parainfluenza virus 2, parainfluenza virus 3, parainfluenza virus 4 in nasopharyngeal (NP) or nasopharyngeal aspirate swabs obtained from individuals exhibiting signs and symptoms of a respiratory tract infection.

Specifications

Content And Storage 5 plates; 3 x 1 mL
Store panels at 15–30°C.
Store master mix at 5 to -30°C.

Detection Method Primer-probe
Label or Dye 2 Color
Quantity 5 Plates
Target Coronavirus 229E, coronavirus HKU1, coronavirus NL63, coronavirus OC43, parainfluenza virus 1, parainfluenza virus 2, parainfluenza virus 3, and parainfluenza virus 4
For Use With (Application) Pathogen Detection
Compatibility QuantStudio™ 5 Real‐Time PCR System, 96-well, 0.2-mL block; QuantStudio™ 5 Dx Real‐Time PCR System, 96-well, 0.2-mL block; QuantStudio™ 7 Pro Real-Time PCR System, 96-well, 0.2-mL block
No. of Reactions Up to 12 Samples/Plate
Does Thermo Fisher Scientific offer analytical validation (AV) consulting services?

When adopting a new testing method for your research needs, we can assist in enabling the verification of the workflow to help ensure a successful launch. Our analytical validation (AV) consulting services can speed up the launch process by up to 75% and cut costs by up to half. Our expert team will consult to help develop and optimize your research workflow, while providing control material, data analysis support, and template documentation to fully maximize your instrument and reagent investment. Please refer to https://www.thermofisher.com/av or contact your sales representative for more information.

What specimen types can be used with the TrueMark Infectious Disease panels?

The recommended specimens that can be used with the TrueMark Infectious Disease panels are listed below:
- TrueMark Respiratory I Plus Combo Kit, TrueMark Respiratory II Plus Combo Kit, and TrueMark Respiratory III Plus Combo Kit: Nasopharyngeal (NP) or nasopharyngeal aspirate swabs
- TrueMark Urinary I Plus Combo Kit, TrueMark Urinary II Plus Combo Kit, and TrueMark Urinary III Plus Combo Kit: Urine samples
- TrueMark STI Plus Combo Kit: Vaginal and genital swabs
- TrueMark Vaginal Plus Combo Kit: Vaginal swabs
- TrueMark Lesion Plus Combo Kit: Lesion/genital swab
- TrueMark Genital Plus Combo Kit: Genital swab

What are the differences between the controls listed in the TrueMark Infectious Disease panels user guide?

Here are the controls listed in the TrueMark Infectious Disease panels user guide and their considerations for use:

TrueMark Amplification Control (Cat. No. A55698) or TrueMark Amplification Control I, low conc. (Cat. No. A55699):
- Added to a single well per target in the real-time PCR plate as a positive control. The recommended concentration for the positive control is 10,000 copies/reaction.
- Monitors real-time PCR reaction setup and reagent integrity.
- Can be ordered separately.
- Includes all the targets in the TrueMark panels.

Negative (extraction) Control:
- It is a negative control on the extraction sample plate that contains water.
- The negative (extraction) control is run on the real-time PCR plate.
- Monitors cross-contamination of reagents during reaction setup.

TaqMan Universal Extraction Control Organism (B. atrophaeus) (Cat. No. A39180):
- Used only for TrueMark Urinary plates.
- Contains three vials of lyophilized Bacillus atrophaeus, for use as a process control for cell lysis and nucleic acid recovery.
- Like other gram-positive bacteria, Bacillus atrophaeus has thick cell walls that can be difficult to lyse. This characteristic makes Bacillus atrophaeus an ideal control to monitor the efficiency of cell lysis and subsequent nucleic acid recovery.
- Is supplied lyophilized with a quantity of 1 x 10E9 copies/vial and is reconstituted in 200 µL of PBS (1X), pH 7.4 to a final concentration of 5 x 10E6 copies/µL.
- The following B. atrophaeus dilution and spike-in process instructions are recommended:
Vortex for 30 seconds, until the pellet is resuspended. Make the suitable number of aliquots and stored at -20 degrees C for up to 4 months. Perform further dilution to obtain the working concentration of 5 x 10E4 copies/µL. 10 µL of the control is added to urine samples in each extraction well at the start of extraction.

What is the approximate workflow time for running the TrueMark Infectious Disease panel assay starting from sample preparation?

It takes approximately 3 hours for running the assay starting from sample preparation to receiving the Ct value.

With TrueMark Infectious Disease panels, how long can we hold the samples before starting the procedure?

We recommend running the plate immediately. If the plate is not ready to run, store it at 2-8 degrees C for up to 2 hours.

What is the recommended negative control for TrueMark Infectious Disease panels?

We recommend using Nuclease-free water as a negative control.

Are there any trial sizes/sample products available for TrueMark Infectious Disease panels?

There are no trial sizes or sample products available.

What is the recommended concentration for the positive amplification control used with TrueMark Infectious Disease panels?

The recommended concentration for the positive amplification control is 10,000 copies/reaction.

What are the recommended positive controls for TrueMark Infectious Disease panels?

- TrueMark Amplification Control I (1 x 10E5 copies/µL), Cat. No. A55699 is the recommended positive control for TrueMark Infectious Disease panels.
- TrueMark Amplification Control I (5 x 10E7 copies/µL), Cat. No. A55698 is recommended for validation purposes.

Do we need to use RNase P with the assays included in the TrueMark Infectious Disease panels?

TrueMark pathogen assays on TaqMan array plates are a collection of duplex assays that detect microbes and an internal control (RNase P) in the same reaction in the RTI and STI panels. This allows for the simultaneous detection of target and internal control in the same well. This also increases confidence in results and reduces the need to run replicates, lowering cost and workflow complexity. Since the RNAse P is dried down in these plates, additional RNAse P is not required to be added to the plates.
Note: Only the RTI and STI panels include RNase P. The internal control for UTI panels (B. atrophaeus) must be purchased separately and added by the user.

What instruments, consumables, and reagents do I need for the TrueMark Infectious Disease panels? Where can I find the full list of products and protocols?

Please download the user guide and lab readiness checklist to see the workflow outlining the instruments, consumables, and reagents required to run TrueMark Infectious Disease panels.

Do you provide run templates for the inventoried TrueMark Infectious Disease panels?

Yes, the run templates can be downloaded from the product page.

What are the components of the TrueMark Infectious Disease panels? Is the master mix included, or does it need to be purchased separately? Are there any other required materials not supplied?

Each inventoried TrueMark Infectious Disease Combo kit includes the following components:
- TrueMark Infectious Disease panel (inventoried TaqMan Array plates): collection of duplex assays that contain primer/probe sets specific to respiratory, urinary, STI pathogens, and RNase P genomic regions
- TrueMark Infectious Disease Panels 1-Step Multiplex Master Mix (No ROX): ready-to-use PCR mix, including reverse transcriptase, polymerase, dNTPs, salt, and buffer
For more details on other consumables, please refer to the user guide or lab readiness checklist.