missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TrueMark™ Enteric Bacterial Select Panel I Combo Kit

Catalog No. A58035
Change view
Click to view available options
Quantity:
200 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A58035 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A58035 Supplier Applied Biosystems™ Supplier No. A58035

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The TrueMark Enteric Bacterial Select Panel I Combo Kit includes the TrueMark Enteric Bacterial Select Panel I and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering.

The TrueMark Enteric Bacterial Select Panel I Combo Kit includes the TrueMark Enteric Bacterial Select Panel I and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering. Together with the TrueMark Enteric Bacterial Select Amplification Control I, the kit can be used to quickly evaluate up to 200 samples and uses extracted nucleic acid from stool specimens. The kit is recommended for use with the QuantStudio 5 Real-Time PCR System (with 96-well, 0.2-mL block). It includes primers and probe sets specific to the following targets:
• Salmonella spp.
• Shigella spp./enteroinvasive E. coli (EIEC)
• Campylobacter jejuni, Campylobacter coli, Campylobacter upsaliensis
• Shiga toxin-producing E. coli (STEC) stx1/stx2
• Bacillus atrophaeus (used as a process control)

Features of the TrueMark Enteric Bacterial Select Panel I Combo Kit include:
• Detects and differentiates between clinically relevant targets found in enteric bacterial infections
• Affordable and scalable—add to existing infrastructure to expand menu while maintaining low operational costs
• Ready-to-use off-the-shelf solution—seamlessly integrate test into the existing workflow

The TrueMark Enteric Bacterial Select Panel I Combo Kit includes the following components:
• TrueMark Enteric Bacterial Select Panel I—multiplexed assay that contains primer and probe sets
• TrueMark Infectious Disease 1‐Step Multiplex Master Mix (No ROX)—ready-to-use PCR mix, including polymerase, dNTPs, salt, and buffer

Use this kit with TrueMark Enteric Bacterial Select Amplification Control I (Cat. No. A58049), which is a plasmid control containing target-specific templates.

Specifications

Detection Method qPCR
Form Liquid
PCR Method qPCR
Reaction Speed 120 min.
Quantity 200 reactions
For Use With (Application) Pathogen Detection
No. of Reactions 200 Reactions