missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Trolox

Catalog No. 361063932
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361063932 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361063932 Supplier R&D Systems™ Supplier No. 6002/100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Antifade reagent; antioxidant vitamin E derivative; cell permeable and water soluble

Trolox is an antioxidant vitamin E derivative; suppresses osteoclast formation and inhibits IL-1-induced osteoclast bone loss. Also inhibits ferroptosis. Commonly used antifade reagent in live cell imaging; suppresses blinking and photobleaching. Cell-permeable and water soluble.

Specifications

Chemical Name or Material 3,4-Dihydro-6-hydroxy-2,5,7,8-tetramethyl-2H-1-benzopyran-2-carboxylic acid
CAS 53188-07-1
Quantity 100 mg
Product Line Tocris
Target Additional Antioxidant Compounds
Molecular Formula C14H18O4
Purity 98%