missing translation for 'onlineSavingsMsg'
Learn More

Trityl Tetrakis(pentafluorophenyl)borate, 97%

Catalog No. p-3213341
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC382390010 1 g Glass bottle
AC382390050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC382390010 Supplier Thermo Scientific Chemicals Supplier No. 382390010
Only null left

CAS: 136040-19-2 | C43H15BF20 | 922.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136040-19-2
Molecular Formula C43H15BF20
Molecular Weight (g/mol) 922.37
MDL Number MFCD03426981
InChI Key TZOSNOQHGGONMD-UHFFFAOYSA-N
Synonym triphenylmethylium tetrakis perfluorophenyl borate, trityl tetrakis pentafluorophenyl borate, trityl tetrapentafluorophenyl borate, methylium, triphenyl-, tetrakis pentafluorophenyl borate 1-, triphenylcarbenium tetrakis pentafluorophenyl borate, methylium, triphenyl-, tetrakis 2,3,4,5,6-pentafluorophenyl borate 1-1:1, tetrakis 2,3,4,5,6-pentafluorophenyl boranuide; triphenylmethylium, methylium, triphenyl-, tetrakis pentafluorophenyl borate 1-1:1, tritylium tetrakis pentafluorophenyl borate, triphenylmethyliumtetrakis pentafluorophenyl borate
PubChem CID 9832824
SMILES C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
IUPAC Name diphenylmethylbenzene;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Formula Weight 922.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Trityl tetrakis(pentafluorophenyl)borate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

RUO – Research Use Only