accessibility menu, dialog, popup

UserName

BFFFFFFFFFFFFFFFFFFFFC

CAS RN 136040-19-2

IUPAC Name: tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide; triphenylmethylium
Synonyms: tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide; tritylium
Molecular Weight (g/mol): 922.37
Molecular Formula: C43H15BF20
InChi Key: TZOSNOQHGGONMD-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)