missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Triton™ X-100, 98%, for molecular biology, DNAse, RNAse and Protease free
CAS: 9002-93-1 | C16H26O2, C16H26O2 | 250.38 g/mol
Supplier: Thermo Scientific Chemicals 327372500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For molecular biologySpecifications
| 98% | |
| 9002-93-1 | |
| 1.0670 g/mL | |
| 646.85 | |
| MFCD00132505 | |
| Glass bottle | |
| 6 to 8 (5% aq. soln.) | |
| 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol | |
| 98%, Molecular Biology | |
| Polyethylene glycol mono [4-(1,1,3,3-tetramethylbutyl)phenyl] ether |
| Triton™ X-100 | |
| Undesignated | |
| 250 mL,250 mL | |
| C16H26O2 | |
| 6.0°C | |
| JYCQQPHGFMYQCF-UHFFFAOYSA-N | |
| CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 | |
| 250.38 | |
| Solubility in water: soluble. Other solubilities: soluble in toluene, xylene, trichloroethylene,, ethylene glycol, ethyl ether, ethyl alcohol,, isopropyl alcohol, ethylene dichloride and most, other solvnets., insoluble in kerosene, miniral spirits. | |
| Viscous Liquid |
Chemical Identifiers
| 9002-93-1 | |
| C16H26O2 | |
| Polyethylene glycol mono [4-(1,1,3,3-tetramethylbutyl)phenyl] ether |
RUO – Research Use Only