missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Triton X-100 (1%)

Catalog No. 501121262
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121262 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121262 Supplier Invitrogen™ Supplier No. HFH10
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Triton X-100 (1%)

This reagent, a component of the Fetal Hemoglobin (HbF) test, is a 0.1% working solution prepared in phosphate-buffered saline (PBS) containing bovine serum albumin (BSA) and sodium azide.

Class: IVD
Statement: For In Vitro Diagnostic Use.
PContains: 0.1% sodium azide
Stabilizer: 0.1% BSA

Specifications

Product Type Detergent
Content And Storage Store in refrigerator (2–8°C).

Shelf life: 24 months from date of manufacture
Shipping Condition Wet Ice
Form Liquid
Reagent Type Triton X-100
Quantity 10 mL

For Research Use Only. Not for use in diagnostic procedures.