missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triton∣r X-100, Electrophoresis Reagent

Catalog No. p-7057520
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66624AE 100 mL
AAJ66624AP 500 mL
AAJ666240F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ66624AE Supplier Thermo Scientific Chemicals Supplier No. J66624AE
Only null left

CAS: 9002-93-1 | C16H26O2, C16H26O2 | 250.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Synonym Octoxynol-9; Polyethylene glycol tert-octylphenyl ether

Specifications

Color Colorless to Yellow
Content And Storage Ambient temperatures
Density 1.061 g/mL
Quantity 100 mL
Formula Weight ca. 625
MDL Number MFCD00132505
Melting Point ∼5°C
Odor Mild
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
Molecular Weight (g/mol) 250.38
Form Liquid
Chemical Name or Material Triton™ X-100
Show More Show Less

RUO – Research Use Only