missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tristearin, Technical grade, ∽65%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210313125
| Quantity | 25 g |
|---|
Chemical Identifiers
| 555-43-1 | |
| 891.501 | |
| tristearin, stearin, glycerol tristearate, glyceryl tristearate, trioctadecanoin, propane-1,2,3-triyl tristearate, hardened oil, stearin, tri, spezialfett 118, stearic triglyceride | |
| CHEBI:45956 | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCC |
| C57H110O6 | |
| DCXXMTOCNZCJGO-UHFFFAOYSA-N | |
| 11146 | |
| 2,3-di(octadecanoyloxy)propyl octadecanoate |
Specifications
| Tristearin | |
| ∽ 65% | |
| 25 g | |
| DCXXMTOCNZCJGO-UHFFFAOYSA-N | |
| 2,3-di(octadecanoyloxy)propyl octadecanoate | |
| 11146 | |
| 891.5 | |
| Crystalline |
| 555-43-1 | |
| C57H110O6 | |
| tristearin, stearin, glycerol tristearate, glyceryl tristearate, trioctadecanoin, propane-1,2,3-triyl tristearate, hardened oil, stearin, tri, spezialfett 118, stearic triglyceride | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCC | |
| 891.501 | |
| CHEBI:45956 | |
| Technical |
Safety and Handling
EINECSNumber : 209-097-6
Recommended Storage : Store at 2° to 8°C.