missing translation for 'onlineSavingsMsg'
Learn More

Tristearin, Technical grade, ∽65%, MP Biomedicals™

Catalog No. p-4404563
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10313105 5 g
ICN10313125 25 g
ICN10313180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10313105 Supplier MP Biomedicals Inc Supplier No. 0210313105

Chemical Identifiers

CAS 555-43-1
Molecular Formula C57H110O6
Molecular Weight (g/mol) 891.501
InChI Key DCXXMTOCNZCJGO-UHFFFAOYSA-N
Synonym tristearin, stearin, glycerol tristearate, glyceryl tristearate, trioctadecanoin, propane-1,2,3-triyl tristearate, hardened oil, stearin, tri, spezialfett 118, stearic triglyceride
PubChem CID 11146
ChEBI CHEBI:45956
IUPAC Name 2,3-di(octadecanoyloxy)propyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCC

Specifications

Assay Percent Range ∽ 65%
Quantity 5 g
Formula Weight 891.5
Grade Technical
Physical Form Crystalline
Chemical Name or Material Tristearin
EINECSNumber 209-097-6
Recommended Storage Store at 2° to 8°C.