accessibility menu, dialog, popup

UserName

H3COOOOCH3OOCH3

CAS RN 555-43-1

IUPAC Name: 1,3-bis(octadecanoyloxy)propan-2-yl octadecanoate
Synonyms: stearin tristearin hydrogenated cottonseed oil
Molecular Weight (g/mol): 891.5
Molecular Formula: C57H110O6
InChi Key: DCXXMTOCNZCJGO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCC
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (3)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)