missing translation for 'onlineSavingsMsg'
Learn More

Tris(triphenylphosphine)ruthenium(II) Dichloride 90.0+%, TCI America™

Catalog No. D19971G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19971G 1 g
D19975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19971G Supplier TCI America Supplier No. D19971G

Chemical Identifiers

CAS 15529-49-4
Molecular Formula C54H45Cl2P3Ru
Molecular Weight (g/mol) 958.85
MDL Number MFCD00013077
InChI Key KSHWMFNJCNGTJO-UHFFFAOYSA-L
Synonym dichlorotris triphenylphosphine ruthenium ii, tris triphenylphosphine ruthenium ii chloride, dichlorotris triphenylphosphino ruthenium ii, tris triphenylphosphine ruthenium ii dichloride, dichlorotris triphenylphosphine ruthenium, ruthenium ii-tris triphenylphosphine dichloride, tris triphenylphosphine dichlororuthenium, ruthenium dichlorotris triphenylphosphine, pubchem12822
PubChem CID 11007548
IUPAC Name λ¹-ruthenium(1+) tris(triphenylphosphane) dichloride
SMILES [Cl-].[Cl-].[Ru+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Black-Green
Quantity 1 g
Formula Weight 958.85
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(triphenylphosphine)ruthenium(II) Dichloride