accessibility menu, dialog, popup

UserName

CAS RN 15529-49-4

IUPAC Name: λ¹-ruthenium(1+) tris(triphenylphosphane) dichloride
Synonyms: λ¹-ruthenium(1+) tris(triphenylphosphine) dichloride
Molecular Weight (g/mol): 958.85
Molecular Formula: C54H45Cl2P3Ru
InChi Key: KSHWMFNJCNGTJO-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Ru+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)