missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS, MP Biomedicals™
TRIS Ultra Pure, ≥99.9%, White crystalline powder
$231.31 - $231.31
Chemical Identifiers
| CAS | 77-86-1 |
|---|---|
| Molecular Formula | C4H11NO3 |
| Molecular Weight (g/mol) | 121.14 |
| MDL Number | MFCD00004679 |
| InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
| Synonym | Tris-[hydroxymethyl]aminomethane, THAM, 2-Amino-2-(hydroxymethyl)- 1,3-propandiol, Tromethamine, Trometamol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MP04819623
|
MP Biomedicals Inc
04819623 |
1 kg |
Each for $231.31
|
|
|||||
Description
- Tris is used in pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.
- Tris is used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions.
- Tris have been useful as buffers in a wide variety of biological systems.
- It has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes).
- It does not precipitate calcium salts and is of value in maintaining solubility of manganese salts.
- It can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid.
- It is RNAse and DNAse-free.Tris is relatively non-hygroscopic;but, if needed, it can be dried at 100°C for up to 4 hours to remove any water.
Chemical Identifiers
| 77-86-1 | |
| 121.14 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N |
| C4H11NO3 | |
| MFCD00004679 | |
| Tris-[hydroxymethyl]aminomethane, THAM, 2-Amino-2-(hydroxymethyl)- 1,3-propandiol, Tromethamine, Trometamol |
Specifications
| 165°C to 175°C | |
| 219°C to 220°C (at 10 mmHg) | |
| 10 to 11.5 (5% aq soln) | |
| 1 kg | |
| C4H11NO3 | |
| 741883 | |
| Soluble in water (550mg/mL), ethyleneglycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL) | |
| 121.14 |
| 77-86-1 | |
| White | |
| Crystalline Powder | |
| ≤1% | |
| MFCD00004679 | |
| Tris-[hydroxymethyl]aminomethane, THAM, 2-Amino-2-(hydroxymethyl)- 1,3-propandiol, Tromethamine, Trometamol | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| ≥99.9% |
Safety and Handling
RTECSNumber : TY2900000
storageNote1 : Room Temperature, desiccate