Learn More
Thermo Scientific™ Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.9%
CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
$109.45 - $395.42
Chemical Identifiers
| CAS | 77-86-1 |
|---|---|
| Molecular Formula | C4H11NO3 |
| Molecular Weight (g/mol) | 121.136 |
| MDL Number | MFCD00004679 |
| InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
| Synonym | 2-Amino-2-hydroxymethyl-1,3-propanediol; TRIS |
| PubChem CID | 6503 |
| ChEBI | CHEBI:9754 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| SMILES | C(C(CO)(CO)N)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6256922
|
Thermo Scientific™
J6256922 |
100 g |
Each for $109.45
|
|
|||||
|
AAJ6256936
|
Thermo Scientific™
J6256936 |
500 g |
Each for $395.42
|
|
|||||
Description
Used as a biological buffer substance in the pH range 7—9; pKa = 8.3 at 20°C; pKa = 7.82 at 37°C. Used as a biological buffer or as a component of buffer formulations such as TAE and TBE buffers. Tris has a pKa of 8.06 and is very useful in the biology and biochemistry lab because it has buffering capabilities in the range of the typical physiological pH of most living organisms (pH 7.0-9.0). Tris has been reported to interfere with the activity of a number of enzymes, it should therefore be used carefully when studying proteins. Used in chemical manufacturing (surface-active agents, vulcanization accelerators, and pharmaceuticals), as an emulsifying agent (cosmetics, mineral oil and paraffin wax emulsions, leather dressings, textile specialties, polishes, cleaning compounds, and soluble oils), and as an absorbent for acidic gases, biological buffer, acidimetric standard, and therapeutic alkalinizing agent. Used in buffer systems for the freezer storage of sperm, to regulate pH in the transport of live aquatic species, cell culture media, and anti-inflammatory drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesStore away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 77-86-1 | |
| 121.136 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| C4H11NO3 | |
| MFCD00004679 | |
| 2-Amino-2-hydroxymethyl-1,3-propanediol; TRIS | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
Specifications
| 168°C to 172°C | |
| 219°C to 220°C (10mmHg) | |
| 1.353 | |
| 100 g | |
| MFCD00004679 | |
| 14,9772 | |
| Not miscible or difficult to mix in water. Soluble in ethylene glycol (79.1mg/ml),methanol (26mg/ml),anhydrous ethanol (14.6mg/ml),95% ethanol (22mg/ml),DMF (14mg/ml),acetone (2mg/ml),ethyl acetate (0.5mg/ml),olive oil (0.4mg/ml),chloroform (0.05mg/ml),and alkaline solution. | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 | |
| 99.9% | |
| Tris(hydroxymethyl)aminomethane |
| 77-86-1 | |
| White | |
| Crystalline Powder | |
| C4H11NO3 | |
| 741883 | |
| 2-Amino-2-hydroxymethyl-1,3-propanediol; TRIS | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| 121.14 | |
| Electrophoresis |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 201-064-4
RTECSNumber : TY2900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only