missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical™
Specifications
Specifications
| Absorbance | Pass Test |
| Melting Point | 168.5°C |
| Boiling Point | 219°C |
| Color | White |
| Physical Form | Powder or Solid |
| Quantity | 1 kg |
| Assay Percent Range | 99.8 to 100.1% |
| Identification | Pass Test |
| Linear Formula | H2NC(CH2OH)3 |
| Packaging | Poly Bottle |
| Show More |