missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical™

Catalog No. T395100
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T395100 100 g Glass Bottle
T395500 500 g Poly Bottle
T3951 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T395100 Supplier Thermo Fisher Scientific Supplier No. FLT395100

Alkalimetric standard

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, Trimethylol Aminomethane, Tris Buffer, Tris, Trizma™, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Absorbance Pass Test
Melting Point 168.5°C
Boiling Point 219°C
Color White
Physical Form Powder or Solid
Quantity 100 g
Assay Percent Range 99.8 to 100.1%
Identification Pass Test
Linear Formula H2NC(CH2OH)3
Packaging Glass Bottle
Formula Weight 121.14
Percent Purity 99.8 to 100.1%
Grade Certified ACS
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less