missing translation for 'onlineSavingsMsg'
Learn More

Tris-Glycine-SDS, 10X Solution (Electrophoresis), Fisher BioReagents™

Catalog No. p-29189
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Nalgene Poly Bottle
Poly CUBE
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP13411 1 L Nalgene Poly Bottle
BP13414 4 L Poly CUBE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP13411 Supplier Fisher BioReagents Supplier No. BP1341 1
Only null left

Tris-Glycine-SDS, 10X Solution (Electrophoresis)

Tris-Glycine-SDS is commonly used as a buffer in SDS-PAGE of proteins.

Specifications

Chemical Name or Material Tris-Glycine-SDS
Name Note 10X Solution
CAS 77-86-1, 7732-18-5, 56-40-6, 151-21-3
Color Undesignated
Physical Form Liquid
Quantity 1 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Purity Grade Notes DNase-, RNase- and Protease-Free
Identification Pass Test
Packaging Nalgene Poly Bottle
Protease Protease free
Synonym TG-SDS
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) 0.25M Tris Base, 1.92M Glycine, and 1.0% (w/v) SDS.
Show More Show Less