missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Tris-Glycine Running Buffer for Native PAGE

Powder buffer salts for making Native-PAGE buffer.

Supplier:  MP Biomedicals™ 08W00011

Catalog No. ICN08W00011


Only null left
Explore available promotions

Description

Description

  • Comes in powder form
  • 1 packet makes up 500 mL
Specifications

Specifications

1X
Native-PAGE
10 Packets
Western Blotting
Powder
Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.