missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TRIS-glycine native sample buffer (4X), pH 8.6
For preparation of protein samples for native gel electrophoresis
$127.10 - $127.10
Specifications
| Color | Blue |
|---|---|
| Concentration | 4X |
| For Use With (Application) | Preparation of protein samples for native gel electrophoresis |
| pH | 8.6 |
| Physical Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ61864AC
|
Thermo Scientific Chemicals
J61864AC |
25 mL |
Each for $127.10
|
|
|||||
Description
For preparation of protein samples for native gel electrophoresis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Blue | |
| Preparation of protein samples for native gel electrophoresis | |
| Liquid |
| 4X | |
| 8.6 |
RUO – Research Use Only