missing translation for 'onlineSavingsMsg'
Learn More

Tris-Glycine, 10X Solution (Electrophoresis), Fisher BioReagents™

Catalog No. BP13064
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Nalgene Poly Bottle
Poly CUBE
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP13064 4 L Poly CUBE
BP13061 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP13064 Supplier Fisher BioReagents Supplier No. BP13064
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tris-Glycine is commonly used as a buffer in native polyacrylamide gel electrophoresis.

Specifications

Chemical Name or Material Tris-Glycine
Name Note 10X Solution
Color Undesignated
CAS 77-86-1, 56-40-6
Physical Form Liquid
Quantity 4 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Purity Grade Notes DNase-, RNase- and Protease-Free
Packaging Poly CUBE
Protease Protease free
Synonym TG
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Components: Tris (2-3%) and Glycine (13-14%).
0.25M Tris Base and 1.92M Glycine.
Show More Show Less