missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, Tris-EDTA, 1X Solution, pH 8.0, Molecular Biology, Fisher BioReagents

Catalog No. p-29171
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2473100 100 mL Poly Bottle
BP2473500 500 mL Poly Bottle
BP24731 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP2473100 Supplier Fisher BioReagents Supplier No. BP2473100
Only null left
  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TBE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 10mM tris and 1mM EDTA
  • Filtered and autoclaved - ready to use
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer (pH 8.0) is routinely used for resuspending DNA after precipitation
  • Intended for laboratory research use only

Specifications

Chemical Name or Material Tris-EDTA
Name Note 1X Solution, pH 8.0
CAS 77-86-1, 6381-92-6, 7647-01-0, 7732-18-5
Color Colorless
pH 7.4 to 8.1
Physical Form Liquid
Quantity 100 mL
DNase DNase free
Filtered Through Filtered through a 5-micron filter.
Packaging Poly Bottle
Protease Protease free
Synonym TE
Grade Molecular Biology
Show More Show Less