missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, Tris-EDTA, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™

Catalog No. p-29148
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2476100 100 mL Poly Bottle
BP2476500 500 mL Poly Bottle
BP24761 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP2476100 Supplier Fisher BioReagents Supplier No. BP2476100
Only null left

Tris-EDTA (TE) is routinely used for suspending nucleic acid samples.

  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 10 mM tris and 1 mM EDTA
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer is routinely used for resuspending nucleic acid samples
  • Intended for laboratory research use only
Emergency Overview: May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA:

  • Health:1
  • Flammability:0
  • Instability:0

Specifications

Chemical Name or Material Tris-EDTA
Name Note 1X Solution, pH 7.4
CAS 77-86-1, 60-00-4, 7647-01-0, 7732-18-5
Color Colorless
pH 7.4 to 8.1
Physical Form Liquid
Quantity 100 mL
DNase DNase free
Filtered Through Filtered through a 5-micron filter.
Packaging Poly Bottle
Protease Protease free
Synonym TE
Grade Molecular Biology
Show More Show Less