missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 10X) pH 7.4

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Supplier:  Thermo Scientific Chemicals J60764K3

 View more versions of this product

Catalog No. AAJ60764K3


Only null left

Description

Description

TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.

Solubility
Soluble in water and organic solvents.

Notes
250mM Tris, 27mM potassium chloride, 1.37M sodium chloride, pH 7.4.
Specifications

Specifications

TRIS-buffered saline
10X
Liquid
C4H11NO3
MFCD00132476
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
77-86-1
7.4
2 L
Odorless
Soluble in water and organic solvents.
C(C(CO)(CO)N)O
121.136
CHEBI:9754

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
MFCD00132476
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only