missing translation for 'onlineSavingsMsg'
Learn More

TBE Buffer, Tris-Borate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents

Catalog No. bp243020
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Nalgene Poly Bottle
Poly CUBE
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP243020 20 L Poly CUBE
BP24301 1 L Nalgene Poly Bottle
BP24304 4 L Poly CUBE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP243020 Supplier Fisher BioReagents Supplier No. BP2430 20
  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TBE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 0.089M tris, 0.089M boric acid, and 0.002M EDTA
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TBE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
  • Intended for laboratory research use only

Specifications

Chemical Name or Material Tris-Borate-EDTA
Name Note 1X Solution
CAS 77-86-1, 6381-92-6, 10043-35-3, 7732-18-5
Color Colorless
pH 8.2 to 8.4
Physical Form Liquid
Quantity 20 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Packaging Poly CUBE
Protease Protease free
Synonym TBE
Grade Electrophoresis
Show More Show Less