missing translation for 'onlineSavingsMsg'
Learn More

Tris Base (White Crystals or Crystalline Powder/Triple-Crystallized), Fisher BioReagents™

For sensitive molecular biology, tissue culture, and electrophoresis methods | CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Supplier:  Fisher BioReagents BP1541

Catalog No. BP154-1



Get Up to $1,800 in Chemicals with Purchase
View all promotions
Only null left

Description

Description

For sensitive molecular biology, tissue culture, and electrophoresis methods

This ultrapure, triple-crystallized buffer component is alcohol-free, so it is compatible with sensitive molecular biology, tissue culture, and electrophoresis methods.
Specifications

Specifications

0.05 max. (1M solution) at 280nm
Alcohol-Free
77-86-1
White
Powder or Solid
DNase free
Pass Test
C4H11NO3
Poly Bottle
15, 9951
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
Tris Base
168.5°C
219°C
10.4
1 kg
DNase-, RNase- and Protease-Free
≥99.9 %
H2NC(CH2OH)3
Protease free
Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
C(C(CO)(CO)N)O
121.136
CHEBI:9754
≥99.9%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions