missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™
DNase-, RNase- and Protease-Free. | CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Supplier: Fisher BioReagents FLBP15210
Description
DNase-, RNase- and Protease-Free.
Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures.Specifications
| 0.05 AU max. at 280nm | |
| 168.5°C | |
| 219°C | |
| 10.4 | |
| 10 kg | |
| DNase-, RNase- and Protease-Free | |
| ≥99.8 % | |
| H2NC(CH2OH)3 | |
| Protease free | |
| Tris(hydroxymethyl)aminomethane, Trimethylol Aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 | |
| ≥99.8% |
| Tris Base | |
| 77-86-1 | |
| White | |
| Powder or Solid | |
| DNase free | |
| Pass Test | |
| C4H11NO3 | |
| Poly Pail | |
| 15, 9951 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| 121.14 | |
| Molecular Biology |
Chemical Identifiers
| 77-86-1 | |
| 121.136 | |
| Tris(hydroxymethyl)aminomethane, Trimethylol Aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
Safety and Handling
Recommended Storage : Room Temperature