missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Tris Base, ULTROL™ Grade, Calbiochem™,
Specifications
Specifications
| Melting Point | 168°C to 172°C |
| Boiling Point | 219°C to 220°C |
| Density | 1.353g/cm3 |
| Quantity | 25 kg |
| Assay Percent Range | ≥99.0% |
| Solubility Information | Soluble in water (800g/L at 20°C) |
| Formula Weight | 121.1 |
| Chemical Name or Material | Tris Base |
| Grade | by titration (dry basis) |
Safety and Handling
| Recommended Storage | 15°C to 30°C (Do not freeze). |