missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Tris Base, ULTROL™ Grade, Calbiochem™,

Catalog No. 6483111KG
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6483111KG 1 kg
6483115KG 5 kg
64831125KG 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6483111KG Supplier MilliporeSigma™ Supplier No. 6483111KG

Useful in the pH range of 10.2 to 10.6

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym tris(Hydroxymethyl)aminomethane
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 168°C to 172°C
Boiling Point 219°C to 220°C
Density 1.353g/cm3
Quantity 1 kg
Assay Percent Range ≥99.0%
Solubility Information Soluble in water (800g/L at 20°C)
Formula Weight 121.1
Chemical Name or Material Tris Base
Grade by titration (dry basis)
Recommended Storage 15°C to 30°C (Do not freeze).