missing translation for 'onlineSavingsMsg'
Learn More

Tris Base, Molecular Grade, Promega™

Numéro de catalogue. prh5133
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PRH5133 100 g
PRH5131 500 g
PRH5135 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. PRH5133 Fournisseur Promega Corporation Code fournisseur H5133
Il en reste null

Commonly used for many molecular biology applications.

  • Quality tested and certified free of DNase, RNase and protease activity

Identifiants chimiques

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Spécifications

Physical Form Crystallized Free Base
Quantity 100 g
Formula Weight 121.14
Grade Molecular Biology
Chemical Name or Material Tris Base
Recommended Storage 15 to 30°C