missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Base, Molecular Grade, Promega™
Commonly used for many molecular biology applications.
Supplier: Promega Corporation H5131
Description
- Quality tested and certified free of DNase, RNase and protease activity
Specifications
| Tris Base | |
| Crystallized Free Base | |
| C4H11NO3 | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 | |
| Molecular Biology |
| 77-86-1 | |
| 500 g | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| 121.14 |
Chemical Identifiers
| 77-86-1 | |
| 121.136 | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
Safety and Handling
Recommended Storage : 15 to 30°C