missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS ACS Ultrapure, Lab Chem
Description
- TRIS ACS Ultrapure Analytical Standard
Specifications
Specifications
| CAS | 77-86-1 |
| For Use With (Application) | Laboratory |
| For Use With (Equipment) | Instruments and Electrodes |
| pH | 10.0 to 11.5 for 5% solution |
| Physical Form | Crystalline Powder |
| Quantity | 5 kg |
| Solution Type | Standard |
| Molecular Weight (g/mol) | 121.135 |
| Percent Purity | 99.8% to 100.2% |
| Grade | ACS |