missing translation for 'onlineSavingsMsg'
Learn More

TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents

Catalog No. p-29045
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1335500 500 mL Poly Bottle
BP13351 1 L Nalgene Poly Bottle
BP13354 4 L Poly CUBE
BP133520 20 L Poly CUBE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BP1335500 Supplier Fisher BioReagents Supplier No. FLBP1335500
Only null left
  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TAE buffer solution - simply dilute as needed
  • 10X solution contains 0.4M tris-acetate and 0.01M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TAE Solution with 9 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 10X Solution
CAS 64-19-7, 7732-18-5, 60-00-4, 77-86-1
Physical Form Liquid
Quantity 500 mL
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Purity Grade Notes DNase-, RNase- and Protease-Free
Packaging Poly Bottle
Protease Protease free
Synonym TAE
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Tris (<5.0%), Acetic Acid (1.0%), and EDTA (<1%).
Show More Show Less