missing translation for 'onlineSavingsMsg'
Learn More

Tris, 99.0 to 101.0% (Dry Basis)

Catalog No. aaj22674a1
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ22674A1 1 kg Plastic bottle
AAJ2267436 500 g Plastic bottle
AAJ22674A7 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ22674A1 Supplier Thermo Fisher Scientific Supplier No. J22674A1
Only null left

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)amino methane
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 171°C to 172°C
Boiling Point 219°C to 220°C (10 mmHg)
Physical Form Powder
Quantity 1 kg
Linear Formula NH2C(CH2OH)3
Packaging Plastic bottle
Beilstein 741883
Formula Weight 121.14
Chemical Name or Material Tris
EINECSNumber 201-064-4
RTECSNumber TY2900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only