missing translation for 'onlineSavingsMsg'
Learn More

Tris, ≥98%

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Supplier:  Thermo Fisher Scientific J22675A1

 View more versions of this product

Catalog No. AAJ22675A1


Only null left

Description

Description

Specifications

Specifications

Tris
77-86-1
Crystalline Powder
C4H11NO3
MFCD00004679
741883
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
171°C to 172°C
219°C to 220°C (10 mmHg)
1 kg
NH2C(CH2OH)3
Plastic bottle
Tris(hydroxymethyl)amino methane
C(C(CO)(CO)N)O
121.136
CHEBI:9754
≥98%

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
C4H11NO3
MFCD00004679
Tris(hydroxymethyl)amino methane
CHEBI:9754
C(C(CO)(CO)N)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-064-4

RTECSNumber : TY2900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only