missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(4-methoxyphenyl)phosphine 97.0+%, TCI America™
Supplier: TCI America T086125G
Specifications
| Tris(4-methoxyphenyl)phosphine | |
| 133°C | |
| C21H21O3P | |
| 25 g | |
| UYUUAUOYLFIRJG-UHFFFAOYSA-N | |
| tris(4-methoxyphenyl)phosphane | |
| 70071 | |
| ≥97.0% (T) |
| 855-38-9 | |
| White-Yellow | |
| MFCD00014896 | |
| tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine | |
| COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC | |
| 352.37 | |
| 352.37 | |
| Crystalline Powder |
Chemical Identifiers
| 855-38-9 | |
| 352.37 | |
| UYUUAUOYLFIRJG-UHFFFAOYSA-N | |
| 70071 | |
| COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC |
| C21H21O3P | |
| MFCD00014896 | |
| tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine | |
| tris(4-methoxyphenyl)phosphane |
Safety and Handling
TSCA : Yes